Learn More
Eurofins Discoverx PathHunter U2OS c-MET/c-MET Dimerization Cell Line, 2 vials. (93-1063C3)
Supplier:  Eurofins Discoverx 931063C3
Encompass
The PathHunter c-MET/c-MET Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. hepatocyte growth factor) induced dimerization of c-MET/c-MET receptors. This product uses EFC technology. The cell line included overexpresses c-MET target protein fused to PK alongside c-MET target protein fused to EA. Upon ligand-induced dimerization of c-MET/c-MET, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1964
Add to Cart
Request a Quote