Learn More
Eurofins Discoverx PathHunter U2OS CSF1R/CSF1R Dimerization Cell Line, 2 vials. (93-1061C3)
Catalog No. 502001961
Encompass
The PathHunter CSF1R/CSF1R Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. M-CSF) induced dimerization of CSF1R/CSF1R receptors. This product uses EFC technology. The cell line included overexpresses CSF1R target protein fused to PK alongside CSF1R target protein fused to EA. Upon ligand-induced dimerization of CSF1R/CSF1R, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1961 Supplier Eurofins Discoverx Supplier No. 931061C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter CSF1R/CSF1R Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. M-CSF) induced dimerization of CSF1R/CSF1R receptors. This product uses EFC technology. The cell line included overexpresses CSF1R target protein fused to PK alongside CSF1R target protein fused to EA. Upon ligand-induced dimerization of CSF1R/CSF1R, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.