Learn More
Eurofins Discoverx PathHunter U2OS CSF2RB/CSF2RA Dimerization Cell Line, 2 vials. (93-1078C3)
Catalog No. 502001989
Encompass
The PathHunter CSF2RB/CSF2RA Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. GM-CSF) induced dimerization of CSF2RB/CSF2RA receptors. This product uses EFC technology. The cell line included overexpresses CSF2RB target protein fused to PK alongside CSF2RA target protein fused to EA. Upon ligand-induced dimerization of CSF2RB/CSF2RA, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1989 Supplier Eurofins Discoverx Supplier No. 931078C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter CSF2RB/CSF2RA Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. GM-CSF) induced dimerization of CSF2RB/CSF2RA receptors. This product uses EFC technology. The cell line included overexpresses CSF2RB target protein fused to PK alongside CSF2RA target protein fused to EA. Upon ligand-induced dimerization of CSF2RB/CSF2RA, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.