Learn More
Eurofins Discoverx PathHunter U2OS CXCR3 β-Arrestin Cell Line, 2 vials. (93-0271C3)
Catalog No. 502000142
Encompass
The PathHunter CXCR3 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CXCL11/I-TAC) based activation of CXCR3 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CXCR3 and EA-tagged β-Arrestin-2. Activation of CXCR3 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0142 Supplier Eurofins Discoverx Supplier No. 930271C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter CXCR3 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CXCL11/I-TAC) based activation of CXCR3 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CXCR3 and EA-tagged β-Arrestin-2. Activation of CXCR3 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.