Learn More
Eurofins Discoverx PathHunter U2OS DRD3 β-Arrestin Cell Line, 2 vials. (93-0591C3)
Catalog No. 502000905
Encompass
The PathHunter DRD3 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. dopamine) based activation of DRD3 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged DRD3 and EA-tagged β-Arrestin-2. Activation of DRD3 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0905 Supplier Eurofins Discoverx Supplier No. 930591C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter DRD3 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. dopamine) based activation of DRD3 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged DRD3 and EA-tagged β-Arrestin-2. Activation of DRD3 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.