Learn More
Eurofins Discoverx PathHunter U2OS EGFR/EGFR Dimerization Cell Line, 2 vials. (93-0989C3)
Catalog No. 502001759
Encompass
The PathHunter EGFR/EGFR Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. EGF) induced dimerization of EGFR/EGFR receptors. This product uses EFC technology. The cell line included overexpresses EGFR target protein fused to PK alongside EGFR target protein fused to EA. Upon ligand-induced dimerization of EGFR/EGFR, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1759 Supplier Eurofins Discoverx Supplier No. 930989C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter EGFR/EGFR Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. EGF) induced dimerization of EGFR/EGFR receptors. This product uses EFC technology. The cell line included overexpresses EGFR target protein fused to PK alongside EGFR target protein fused to EA. Upon ligand-induced dimerization of EGFR/EGFR, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.