Learn More
Eurofins Discoverx PathHunter U2OS EGFR/ErbB3 Dimerization Cell Line, 2 vials. (93-1125C3)
Supplier:  Eurofins Discoverx 931125C3
Encompass
The PathHunter EGFR/ErbB3 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. heregulinβ-1) induced dimerization of EGFR/ErbB3 receptors. This product uses EFC technology. The cell line included overexpresses ErbB3 target protein fused to PK alongside EGFR target protein fused to EA. Upon ligand-induced dimerization of EGFR/ErbB3, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-2145
Only null left
Add to Cart
Request a Quote