Learn More
Eurofins Discoverx PathHunter U2OS EpoR/EpoR Dimerization Cell Line, 2 vials. (93-0965C3)
Catalog No. 502001692
Encompass
The PathHunter EpoR/EpoR Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. erythropoietin) induced dimerization of EpoR/EpoR receptors. This product uses EFC technology. The cell line included overexpresses EpoR target protein fused to PK alongside EpoR target protein fused to EA. Upon ligand-induced dimerization of EpoR/EpoR, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.
Catalog No. 50-200-1692 Supplier Eurofins Discoverx Supplier No. 930965C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter EpoR/EpoR Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. erythropoietin) induced dimerization of EpoR/EpoR receptors. This product uses EFC technology. The cell line included overexpresses EpoR target protein fused to PK alongside EpoR target protein fused to EA. Upon ligand-induced dimerization of EpoR/EpoR, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.