Learn More
Eurofins Discoverx PathHunter U2OS ErbB2/ErbB4 Dimerization Cell Line, 2 vials. (93-0960C3)
Supplier:  Eurofins Discoverx 930960C3
Encompass
The PathHunter ErbB2/ErbB4 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. heregulin β-1) induced dimerization of ErbB2/ErbB4 receptors. This product uses EFC technology. The cell line included overexpresses ErbB2 target protein fused to PK alongside ErbB4 target protein fused to EA. Upon ligand-induced dimerization of ErbB2/ErbB4, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1673
Add to Cart
Request a Quote