Learn More
Eurofins Discoverx PathHunter U2OS F2RL1 (PAR2, GPR11) β-Arrestin Cell Line, 2 vials. (93-0235C3)
Supplier:  Eurofins Discoverx 930235C3
Encompass
The PathHunter F2RL1 (PAR2,GPR11) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. proteases, SLIGRL) based activation of F2RL1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged F2RL1 and EA-tagged β-Arrestin-2. Activation of F2RL1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0027
Only null left
Add to Cart
Request a Quote