Learn More
Eurofins Discoverx PathHunter U2OS FGFR1-β-Klotho Functional Assay, 2 vials. (93-1118C3)
Catalog No. 502002139
Encompass
The PathHunter U2OS FGFR1-β-Klotho Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated FGFR1 and β-Klotho dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses ProLink-tagged FGFR1 and an EA-tagged SH2 domain protein. It also overexpresses an untagged coreceptor, β-Klotho. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-2139 Supplier Eurofins Discoverx Supplier No. 931118C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter U2OS FGFR1-β-Klotho Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated FGFR1 and β-Klotho dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses ProLink-tagged FGFR1 and an EA-tagged SH2 domain protein. It also overexpresses an untagged coreceptor, β-Klotho. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.