Learn More
Eurofins Discoverx PathHunter U2OS FGFR3/FGFR3 Dimerization Cell Line, 2 vials. (93-1055C3)
Catalog No. 502001949
Encompass
The PathHunter FGFR3/FGFR3 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. FGF-18) induced dimerization of FGFR3/FGFR3 receptors. This product uses EFC technology. The cell line included overexpresses FGFR3 target protein fused to PK alongside FGFR3 target protein fused to EA. Upon ligand-induced dimerization of FGFR3/FGFR3, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1949 Supplier Eurofins Discoverx Supplier No. 931055C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter FGFR3/FGFR3 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. FGF-18) induced dimerization of FGFR3/FGFR3 receptors. This product uses EFC technology. The cell line included overexpresses FGFR3 target protein fused to PK alongside FGFR3 target protein fused to EA. Upon ligand-induced dimerization of FGFR3/FGFR3, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.