Learn More
Eurofins Discoverx PathHunter U2OS FLT4 Activity Assay, 2 vials. (93-0888C3)
Catalog No. 502001506
Encompass
The PathHunter FLT4 RTK Activity cell line is a stable, engineered cell line for use in studying small molecule drug candidates targeting RTKs. This cell line enables assessment of RTK activity via detection of resulting FLT4 phosphorylation-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses ProLink-tagged FLT4 to the extracellular ligand binding domain of EGFR alongside an EA-tagged SH2 domain protein. Upon ligand induced receptor activation, cross-phosphorylation of the receptor leads to SH2-EA recruitment and production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1506 Supplier Eurofins Discoverx Supplier No. 930888C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter FLT4 RTK Activity cell line is a stable, engineered cell line for use in studying small molecule drug candidates targeting RTKs. This cell line enables assessment of RTK activity via detection of resulting FLT4 phosphorylation-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses ProLink-tagged FLT4 to the extracellular ligand binding domain of EGFR alongside an EA-tagged SH2 domain protein. Upon ligand induced receptor activation, cross-phosphorylation of the receptor leads to SH2-EA recruitment and production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.