Learn More
Eurofins Discoverx PathHunter U2OS FOXO3 Nuclear Translocation Cell Line, 2 vials. (93-0876C3)
Catalog No. 502001472
Encompass
The PathHunter U2OS FOXO3 Nuclear Translocation Cell Line is a stable, engineered cell line for use in studying drug candidates affecting the PI3 pathway. This cell line enables assessment of ligand (e.g. wortmannin) based pathway activation via detection of resulting FOXO3 nuclear translocation. This product uses EFC technology. The included cell line overexpresses ProLabel-tagged FOXO3 and nuclear localized EA. Upon pathway activation, induced nuclear translocation of FOXO3 leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/pathways.
Catalog No. 50-200-1472 Supplier Eurofins Discoverx Supplier No. 930876C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter U2OS FOXO3 Nuclear Translocation Cell Line is a stable, engineered cell line for use in studying drug candidates affecting the PI3 pathway. This cell line enables assessment of ligand (e.g. wortmannin) based pathway activation via detection of resulting FOXO3 nuclear translocation. This product uses EFC technology. The included cell line overexpresses ProLabel-tagged FOXO3 and nuclear localized EA. Upon pathway activation, induced nuclear translocation of FOXO3 leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/pathways.