Learn More
Eurofins Discoverx PathHunter U2OS GHSR1a β-Arrestin-1 Cell Line, 2 vials. (93-0242C3)
Catalog No. 502000055
Encompass
The PathHunter GHSR1a (ARR1) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. ghrelin) based activation of GHSR1a GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GHSR1a and EA-tagged β-Arrestin-1. Activation of GHSR1a stimulates the recruitment of β-Arrestin-1 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0055 Supplier Eurofins Discoverx Supplier No. 930242C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter GHSR1a (ARR1) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. ghrelin) based activation of GHSR1a GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GHSR1a and EA-tagged β-Arrestin-1. Activation of GHSR1a stimulates the recruitment of β-Arrestin-1 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.