Learn More
Eurofins Discoverx PathHunter U2OS GPBAR1 β-Arrestin Cell Line, 2 vials. (93-0794C3)
Supplier:  Eurofins Discoverx 930794C3
Encompass
The PathHunter GPBAR1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. bile acids) based activation of GPBAR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPBAR1 and EA-tagged β-Arrestin-2. Activation of GPBAR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1318
Add to Cart
Request a Quote