Learn More
Eurofins Discoverx PathHunter U2OS GPR22 β-Arrestin Orphan GPCR Cell Line, 2 vials. (93-1085C3)
Supplier:  Eurofins Discoverx 931085C3
Encompass
The PathHunter GPR22 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand based activation of GPR22 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPR22 and EA-tagged β-Arrestin-2. Activation of GPR22 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. This is an orphan receptor without a known ligand. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-2014
Add to Cart
Request a Quote