Learn More
Eurofins Discoverx U2OS GPR39 Gq Cell Line, 2 vials. (95-1015C3)
Catalog No. 502004090
Encompass
The GPR39 Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. Zn2+) based activation of GPR39 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses GPR39, a naturally Gq coupled GPCR. Upon activation of GPR39, the resulting calcium activity can be fluorescently assayed using the Calcium No WashPLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-4090 Supplier Eurofins Discoverx Supplier No. 951015C3
Add to Cart
Add to Cart
Request a Quote
The GPR39 Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. Zn2+) based activation of GPR39 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses GPR39, a naturally Gq coupled GPCR. Upon activation of GPR39, the resulting calcium activity can be fluorescently assayed using the Calcium No WashPLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.