Learn More
Eurofins Discoverx PathHunter U2OS GPR52 β-Arrestin Orphan GPCR Cell Line, 2 vials. (93-0371C3)
Catalog No. 502000376
Encompass
The PathHunter GPR52 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand based activation of GPR52 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPR52 and EA-tagged β-Arrestin-2. Activation of GPR52 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. This is an orphan receptor without a known ligand. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0376 Supplier Eurofins Discoverx Supplier No. 930371C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter GPR52 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand based activation of GPR52 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPR52 and EA-tagged β-Arrestin-2. Activation of GPR52 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. This is an orphan receptor without a known ligand. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.