Learn More
Eurofins Discoverx PathHunter U2OS GRPR β-Arrestin Cell Line, 2 vials. (93-0229C3)
Catalog No. 502000006
Encompass
The PathHunter GRPR cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. gastrin-releasing peptide) based activation of GRPR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GRPR and EA-tagged β-Arrestin-2. Activation of GRPR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0006 Supplier Eurofins Discoverx Supplier No. 930229C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter GRPR cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. gastrin-releasing peptide) based activation of GRPR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GRPR and EA-tagged β-Arrestin-2. Activation of GRPR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.