Learn More
Eurofins Discoverx PathHunter U2OS HRH4 β-Arrestin Cell Line, 2 vials. (93-0209C3)
Supplier:  Eurofins Discoverx 930209C3
Encompass
The PathHunter HRH4 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. histamine) based activation of HRH4 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged HRH4 and EA-tagged β-Arrestin-2. Activation of HRH4 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-199-9957
Add to Cart
Request a Quote