Learn More
Eurofins Discoverx PathHunter U2OS HTR1B β-Arrestin Cell Line, 2 vials. (93-0697C3)
Catalog No. 502001139
Encompass
The PathHunter HTR1B cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin) based activation of HTR1B GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged HTR1B and EA-tagged β-Arrestin-2. Activation of HTR1B stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1139 Supplier Eurofins Discoverx Supplier No. 930697C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter HTR1B cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin) based activation of HTR1B GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged HTR1B and EA-tagged β-Arrestin-2. Activation of HTR1B stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.