Learn More
Eurofins Discoverx PathHunter U2OS HTR1E β-Arrestin Cell Line, 2 vials. (93-1030C3)
Catalog No. 502001884
Encompass
The PathHunter HTR1E cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin) based activation of HTR1E GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged HTR1E and EA-tagged β-Arrestin-2. Activation of HTR1E stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1884 Supplier Eurofins Discoverx Supplier No. 931030C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter HTR1E cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin) based activation of HTR1E GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged HTR1E and EA-tagged β-Arrestin-2. Activation of HTR1E stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.