Learn More
Eurofins Discoverx PathHunter U2OS HTR1F β-Arrestin Cell Line, 2 vials. (93-0699C3)
Supplier:  Eurofins Discoverx 930699C3
Encompass
The PathHunter HTR1F cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin) based activation of HTR1F GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged HTR1F and EA-tagged β-Arrestin-2. Activation of HTR1F stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1143
Add to Cart
Request a Quote