Learn More
Eurofins Discoverx PathHunter U2OS HTR2C (VGV) β-Arrestin Cell Line, 2 vials. (93-0932C3)
Catalog No. 502001596
Encompass
The PathHunter HTR2C(VGV) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin) based activation of HTR2C(VGV) GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged HTR2C(VGV) and EA-tagged β-Arrestin-2. Activation of HTR2C(VGV) stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1596 Supplier Eurofins Discoverx Supplier No. 930932C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter HTR2C(VGV) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin) based activation of HTR2C(VGV) GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged HTR2C(VGV) and EA-tagged β-Arrestin-2. Activation of HTR2C(VGV) stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.