Learn More
Eurofins Discoverx PathHunter U2OS HTR5A β-Arrestin Cell Line, 2 vials. (93-0700C3)
Catalog No. 502001147
Encompass
The PathHunter HTR5A cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin) based activation of HTR5A GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged HTR5A and EA-tagged β-Arrestin-2. Activation of HTR5A stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1147 Supplier Eurofins Discoverx Supplier No. 930700C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter HTR5A cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin) based activation of HTR5A GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged HTR5A and EA-tagged β-Arrestin-2. Activation of HTR5A stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.