Learn More
Eurofins Discoverx cAMP Hunter U2OS HTR7B Gs Cell Line, 2 vials. (95-0156C3)
Catalog No. 502003842
Encompass
The cAMP Hunter HTR7B Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin/5-HT) based activation of HTR7B GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type HTR7B, a naturally Gs coupled GPCR. Upon activation of HTR7B, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-3842 Supplier Eurofins Discoverx Supplier No. 950156C3
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter HTR7B Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin/5-HT) based activation of HTR7B GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type HTR7B, a naturally Gs coupled GPCR. Upon activation of HTR7B, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.