Learn More
Eurofins Discoverx PathHunter U2OS IFNGR1/IFNGR2 Dimerization Cell Line, 2 vials. (93-1098C3)
Supplier:  Eurofins Discoverx 931098C3
Encompass
The PathHunter IFNGR1/IFNGR2 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IFN-g) induced dimerization of IFNGR1/IFNGR2 receptors. This product uses EFC technology. The cell line included overexpresses IFNGR1 target protein fused to PK alongside IFNGR2 target protein fused to EA. Upon ligand-induced dimerization of IFNGR1/IFNGR2, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.
Catalog No. 50-200-2068
Only null left
Add to Cart
Request a Quote