Learn More
Eurofins Discoverx PathHunter U2OS IFNRL1/IL10RB Dimerization Cell Line, 2 vials. (93-0993C3)
Catalog No. 502001765
Encompass
The PathHunter IFNRL1/IL10RB Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-29) induced dimerization of IFNRL1/IL10RB receptors. This product uses EFC technology. The cell line included overexpresses IFNRL1 target protein fused to PK alongside IL10RB target protein fused to EA. Upon ligand-induced dimerization of IFNRL1/IL10RB, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.
Catalog No. 50-200-1765 Supplier Eurofins Discoverx Supplier No. 930993C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter IFNRL1/IL10RB Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-29) induced dimerization of IFNRL1/IL10RB receptors. This product uses EFC technology. The cell line included overexpresses IFNRL1 target protein fused to PK alongside IL10RB target protein fused to EA. Upon ligand-induced dimerization of IFNRL1/IL10RB, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.