Learn More
Eurofins Discoverx PathHunter U2OS IL17RD/TNFR2 Dimerization Cell Line, 2 vials. (93-1068C3)
Catalog No. 502001973
Encompass
The PathHunter IL17RD/TNFR2  Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. TNFα) induced dimerization of IL17RD/TNFR2  receptors. This product uses EFC technology. The cell line included overexpresses IL17RD target protein fused to PK alongside TNFR2 target protein fused to EA. Upon ligand-induced dimerization of IL17RD/TNFR2 , detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.
Catalog No. 50-200-1973 Supplier Eurofins Discoverx Supplier No. 931068C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter IL17RD/TNFR2  Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. TNFα) induced dimerization of IL17RD/TNFR2  receptors. This product uses EFC technology. The cell line included overexpresses IL17RD target protein fused to PK alongside TNFR2 target protein fused to EA. Upon ligand-induced dimerization of IL17RD/TNFR2 , detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.