Learn More
Eurofins Discoverx PathHunter U2OS IL2RB/IL2RG/IL2RA Dimerization Cell Line, 2 vials. (93-1003C3)
Catalog No. 502001811
Encompass
The PathHunter IL2RB/IL2RG/IL2RA Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-2) induced dimerization of IL2RB/IL2RG/IL2RA receptors. This product uses EFC technology. The cell line included overexpresses IL2RB target protein fused to PK alongside IL2RG target protein fused to EA and untagged ILR2A. Upon ligand-induced dimerization of IL2RB/IL2RG/IL2RA, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.
Catalog No. 50-200-1811 Supplier Eurofins Discoverx Supplier No. 931003C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter IL2RB/IL2RG/IL2RA Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-2) induced dimerization of IL2RB/IL2RG/IL2RA receptors. This product uses EFC technology. The cell line included overexpresses IL2RB target protein fused to PK alongside IL2RG target protein fused to EA and untagged ILR2A. Upon ligand-induced dimerization of IL2RB/IL2RG/IL2RA, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.