Learn More
Eurofins Discoverx PathHunter U2OS IL4R/IL13RA1 Dimerization Cell Line, 2 vials. (93-1000C3)
Catalog No. 502001795
Encompass
The PathHunter IL4R/IL13RA1 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-13) induced dimerization of IL4R/IL13RA1 receptors. This product uses EFC technology. The cell line included overexpresses IL4R target protein fused to PK alongside IL13RA1 target protein fused to EA. Upon ligand-induced dimerization of IL4R/IL13RA1, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.
Catalog No. 50-200-1795 Supplier Eurofins Discoverx Supplier No. 931000C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter IL4R/IL13RA1 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-13) induced dimerization of IL4R/IL13RA1 receptors. This product uses EFC technology. The cell line included overexpresses IL4R target protein fused to PK alongside IL13RA1 target protein fused to EA. Upon ligand-induced dimerization of IL4R/IL13RA1, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.