Learn More
Eurofins Discoverx PathHunter U2OS IL4R/IL2RG Dimerization Cell Line, 2 vials. (93-0988C3)
Supplier:  Eurofins Discoverx 930988C3
Encompass
The PathHunter IL4R/IL2RG Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-4) induced dimerization of IL4R/IL2RG receptors. This product uses EFC technology. The cell line included overexpresses IL4R target protein fused to PK alongside IL2RG target protein fused to EA. Upon ligand-induced dimerization of IL4R/IL2RG, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.
Catalog No. 50-200-1755
Add to Cart
Request a Quote