Learn More
Eurofins Discoverx PathHunter U2OS JAK3 Activity Assay, 2 vials. (93-0796C3)
Catalog No. 502001319
Encompass
The PathHunter U2OS JAK3 Activity Assay is a stable, engineered cell line for use in studying small molecule drug candidates targeting the JAK3 kinase domain. The cell line is engineered to express the catalytic domain from JAK3 fused to the cytokine receptor binding domain (CRBD) of JAK2. The cells co-express an Enzyme Acceptor (EA) tagged SH2 domain and a ProLink (PK) tagged Cytokine receptor. The CRBD is designed to bind the CTK catalytic domain to the PRLR-PK. Activation of the PRLR-PK induces receptor phosphorylation by the JAK3 catalytic domain. This leads to SH2-EA recruitment and production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1319 Supplier Eurofins Discoverx Supplier No. 930796C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter U2OS JAK3 Activity Assay is a stable, engineered cell line for use in studying small molecule drug candidates targeting the JAK3 kinase domain. The cell line is engineered to express the catalytic domain from JAK3 fused to the cytokine receptor binding domain (CRBD) of JAK2. The cells co-express an Enzyme Acceptor (EA) tagged SH2 domain and a ProLink (PK) tagged Cytokine receptor. The CRBD is designed to bind the CTK catalytic domain to the PRLR-PK. Activation of the PRLR-PK induces receptor phosphorylation by the JAK3 catalytic domain. This leads to SH2-EA recruitment and production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.