Learn More
Eurofins Discoverx PathHunter U2OS KISS1R β-Arrestin Cell Line, 2 vials. (93-0927C3)
Catalog No. 502001580
Encompass
The PathHunter KISS1R cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. kisspeptin) based activation of KISS1R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged KISS1R and EA-tagged β-Arrestin-2. Activation of KISS1R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1580 Supplier Eurofins Discoverx Supplier No. 930927C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter KISS1R cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. kisspeptin) based activation of KISS1R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged KISS1R and EA-tagged β-Arrestin-2. Activation of KISS1R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.