Learn More
Eurofins Discoverx PathHunter U2OS LEPR/LEPR Dimerization Cell Line, 2 vials. (93-1107C3)
Supplier:  Eurofins Discoverx 931107C3
Encompass
The PathHunter LEPR/LEPR Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. leptin) induced dimerization of LEPR/LEPR receptors. This product uses EFC technology. The cell line included overexpresses LEPR target protein fused to PK alongside LEPR target protein fused to EA. Upon ligand-induced dimerization of LEPR/LEPR, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-2120
Add to Cart
Request a Quote