Learn More
Eurofins Discoverx PathHunter U2OS MC4R β-Arrestin Cell Line, 2 vials. (93-0211C3)
Catalog No. 501999961
Encompass
The PathHunter MC4R cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. α-MSH) based activation of MC4R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged MC4R and EA-tagged β-Arrestin-2. Activation of MC4R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-199-9961 Supplier Eurofins Discoverx Supplier No. 930211C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter MC4R cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. α-MSH) based activation of MC4R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged MC4R and EA-tagged β-Arrestin-2. Activation of MC4R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.