Learn More
Eurofins Discoverx PathHunter U2OS MCHR2 β-Arrestin Cell Line, 2 vials. (93-0691C3)
Catalog No. 502001119
Encompass
The PathHunter MCHR2 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. MCH) based activation of MCHR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged MCHR2 and EA-tagged β-Arrestin-2. Activation of MCHR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1119 Supplier Eurofins Discoverx Supplier No. 930691C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter MCHR2 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. MCH) based activation of MCHR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged MCHR2 and EA-tagged β-Arrestin-2. Activation of MCHR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.