Learn More
Eurofins Discoverx PathHunter U2OS mCXCR7 β-Arrestin Cell Line, 2 vials. (93-0689C3)
Catalog No. 502001111
Encompass
The PathHunter mCXCR7 (mCMKOR1) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CXCL12/SDF-1α) based activation of mCXCR7 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mCXCR7 and EA-tagged β-Arrestin-2. Activation of mCXCR7 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1111 Supplier Eurofins Discoverx Supplier No. 930689C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter mCXCR7 (mCMKOR1) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CXCL12/SDF-1α) based activation of mCXCR7 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mCXCR7 and EA-tagged β-Arrestin-2. Activation of mCXCR7 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.