Learn More
Eurofins Discoverx PathHunter U2OS mGIPR β-Arrestin Cell Line, 2 vials. (93-0846C3)
Catalog No. 502001412
Encompass
The PathHunter mGIPR cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. GIP) based activation of mGIPR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mGIPR and EA-tagged β-Arrestin-2. Activation of mGIPR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1412 Supplier Eurofins Discoverx Supplier No. 930846C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter mGIPR cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. GIP) based activation of mGIPR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mGIPR and EA-tagged β-Arrestin-2. Activation of mGIPR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.