Learn More
Eurofins Discoverx PathHunter U2OS mNPY1R β-Arrestin Cell Line, 2 vials. (93-0610C3)
Catalog No. 502000938
Encompass
The PathHunter mNPY1R cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. peptide YY) based activation of mNPY1R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mNPY1R and EA-tagged β-Arrestin-2. Activation of mNPY1R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0938 Supplier Eurofins Discoverx Supplier No. 930610C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter mNPY1R cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. peptide YY) based activation of mNPY1R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mNPY1R and EA-tagged β-Arrestin-2. Activation of mNPY1R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.