Learn More
Eurofins Discoverx PathHunter U2OS mPD-L1 Ligand Cell Line, 2 vials. (93-1159C3)
Catalog No. 502002275
Encompass
The PathHunter mPD-L1 Ligand cell line is a stable, engineered cell line designed as a companion product for use with mPD-1 Checkpoint Signaling Cell Lines. When co-cultured, the mPD-L1 expressed in this cell line will induce activity of mPD-1 receptors, which may be assayed through SH2 domain protein recruitment. The cell line included overexpresses mPD-L1 cell surface ligand. When co-cultured with the mPD-1 Checkpoint Signaling Cell Line, the mPD-L1 ligand binds to the checkpoint receptor, inducing receptor activity. This receptor activity leads to recruitment of the relevant SH2 domain protein to the checkpoint receptor inside the receptor cell line and production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/checkpoint.
Catalog No. 50-200-2275 Supplier Eurofins Discoverx Supplier No. 931159C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter mPD-L1 Ligand cell line is a stable, engineered cell line designed as a companion product for use with mPD-1 Checkpoint Signaling Cell Lines. When co-cultured, the mPD-L1 expressed in this cell line will induce activity of mPD-1 receptors, which may be assayed through SH2 domain protein recruitment. The cell line included overexpresses mPD-L1 cell surface ligand. When co-cultured with the mPD-1 Checkpoint Signaling Cell Line, the mPD-L1 ligand binds to the checkpoint receptor, inducing receptor activity. This receptor activity leads to recruitment of the relevant SH2 domain protein to the checkpoint receptor inside the receptor cell line and production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/checkpoint.