Learn More
Eurofins Discoverx PathHunter U2OS mRHO(P23H) MEM-EA Pharmacotrafficking Assay, 2 vials. (93-1077C3)
Catalog No. 502001988
Encompass
The PathHunter U2OS mRHO(P23H) MEM-EA Pharmacotrafficking Assay is a stable, engineered cell line for use in studying pharmacochaperones targeting mRHO. This cell line enables assessment of chaperone-based stabilization of mRHO(P23H) via detection of rescued membrane trafficking. The included cell line overexpresses ProLink-tagged mutant mRHO(P23H), which is misfolded and retained in the ER. The cell line also expresses membrane-localized EA. When a pharmacochaperone stabilizes the mutant mRHO, it is trafficked to the membrane, producing EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1988 Supplier Eurofins Discoverx Supplier No. 931077C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter U2OS mRHO(P23H) MEM-EA Pharmacotrafficking Assay is a stable, engineered cell line for use in studying pharmacochaperones targeting mRHO. This cell line enables assessment of chaperone-based stabilization of mRHO(P23H) via detection of rescued membrane trafficking. The included cell line overexpresses ProLink-tagged mutant mRHO(P23H), which is misfolded and retained in the ER. The cell line also expresses membrane-localized EA. When a pharmacochaperone stabilizes the mutant mRHO, it is trafficked to the membrane, producing EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.