Learn More
Eurofins Discoverx PathHunter U2OS NMU1R β-Arrestin Cell Line, 2 vials. (93-0232C3)
Supplier:  Eurofins Discoverx 930232C3
Encompass
The PathHunter nMU1R cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. neuromedin U) based activation of nMU1R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged nMU1R and EA-tagged β-Arrestin-2. Activation of nMU1R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0014
Add to Cart
Request a Quote