Learn More
Eurofins Discoverx PathHunter U2OS NPSR1 β-Arrestin Cell Line, 2 vials. (93-0822C3)
Catalog No. 502001359
Encompass
The PathHunter NPSR1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. neuropeptide S) based activation of NPSR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged NPSR1 and EA-tagged β-Arrestin-2. Activation of NPSR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1359 Supplier Eurofins Discoverx Supplier No. 930822C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter NPSR1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. neuropeptide S) based activation of NPSR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged NPSR1 and EA-tagged β-Arrestin-2. Activation of NPSR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.