Learn More
Eurofins Discoverx PathHunter U2OS OPRM1 β-Arrestin Cell Line, 2 vials. (93-0213C3)
Catalog No. 501999972
Encompass
The PathHunter OPRM1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. met 5-enkephalin) based activation of OPRM1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged OPRM1 and EA-tagged β-Arrestin-2. Activation of OPRM1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-199-9972 Supplier Eurofins Discoverx Supplier No. 930213C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter OPRM1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. met 5-enkephalin) based activation of OPRM1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged OPRM1 and EA-tagged β-Arrestin-2. Activation of OPRM1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.