Learn More
Eurofins Discoverx PathHunter U2OS OSMR/IL6ST Dimerization Cell Line, 2 vials. (93-1056C3)
Catalog No. 502001950
Encompass
The PathHunter OSMR/IL6ST Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. oncostatin M) induced dimerization of OSMR/IL6ST receptors. This product uses EFC technology. The cell line included overexpresses IL6ST target protein fused to PK alongside OSMR target protein fused to EA. Upon ligand-induced dimerization of OSMR/IL6ST, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.
Catalog No. 50-200-1950 Supplier Eurofins Discoverx Supplier No. 931056C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter OSMR/IL6ST Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. oncostatin M) induced dimerization of OSMR/IL6ST receptors. This product uses EFC technology. The cell line included overexpresses IL6ST target protein fused to PK alongside OSMR target protein fused to EA. Upon ligand-induced dimerization of OSMR/IL6ST, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.