Learn More
Eurofins Discoverx PathHunter U2OS OSMRb/IL31R Dimerization Cell Line, 2 vials. (93-1002C3)
Catalog No. 502001803
Encompass
The PathHunter OSMRb/IL31R Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-31) induced dimerization of OSMRb/IL31R receptors. This product uses EFC technology. The cell line included overexpresses OSMRb target protein fused to PK alongside IL31RA target protein fused to EA. Upon ligand-induced dimerization of OSMRb/IL31R, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.
Catalog No. 50-200-1803 Supplier Eurofins Discoverx Supplier No. 931002C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter OSMRb/IL31R Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-31) induced dimerization of OSMRb/IL31R receptors. This product uses EFC technology. The cell line included overexpresses OSMRb target protein fused to PK alongside IL31RA target protein fused to EA. Upon ligand-induced dimerization of OSMRb/IL31R, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.