Learn More
Eurofins Discoverx PathHunter U2OS OX40 Signaling Assay, 2 vials. (93-1080C3)
Catalog No. 502001999
Encompass
The PathHunter OX40 Checkpoint Signaling cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting signaling checkpoints. This cell line enables assessment of OX40 induced non-canonical NF-κB signaling via detection of NIK stabilization. This product uses EFC technology. The included cell line overexpresses ED-tagged NIK (NF-kappa-B Inducing Kinase) protein as well as untagged OX40 receptor. Upon activation of the receptor by its ligand or an antibody, NIK is stabilized, allowing detection of EFC signal when treated with exogenous EA. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/checkpoint.
Catalog No. 50-200-1999 Supplier Eurofins Discoverx Supplier No. 931080C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter OX40 Checkpoint Signaling cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting signaling checkpoints. This cell line enables assessment of OX40 induced non-canonical NF-κB signaling via detection of NIK stabilization. This product uses EFC technology. The included cell line overexpresses ED-tagged NIK (NF-kappa-B Inducing Kinase) protein as well as untagged OX40 receptor. Upon activation of the receptor by its ligand or an antibody, NIK is stabilized, allowing detection of EFC signal when treated with exogenous EA. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/checkpoint.