Learn More
Eurofins Discoverx PathHunter U2OS P2RY1 β-Arrestin Cell Line, 2 vials. (93-0941C3)
Catalog No. 502001624
Encompass
The PathHunter P2RY1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. ATP) based activation of P2RY1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged P2RY1 and EA-tagged β-Arrestin-2. Activation of P2RY1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1624 Supplier Eurofins Discoverx Supplier No. 930941C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter P2RY1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. ATP) based activation of P2RY1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged P2RY1 and EA-tagged β-Arrestin-2. Activation of P2RY1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.